ContactUsLinkComponent
Recombinant STIM1 produced in E.coli is a single, non-glycosylated polypeptide containing 343 amino acids and having a molecular ma ss of 38 kDa. STIM1 is fused to Calmodulin and purified by conventional chromatography techniques. 1 * 25 µG
Catalog # BIRBORB81389-25
Supplier:  Biorbyt
Recombinant STIM1 produced in E.coli is a single, non-glycosylated polypeptide containing 343 amino acids and having a molecular ma ss of 38 kDa. STIM1 is fused to Calmodulin and purified by conventional chromatography techniques. 1 * 25 µG
Catalog # BIRBORB81389-25
Supplier:  Biorbyt