sRAGE Human Recombinant produced in HEK cells is a single, glycosylated, polypeptide chain containing 331 amino acids and having a m olecular mass of 35.2 kDa. sRAGE Human is fused to Flag tag at C-Terminus.sRAGE Human is purified by proprie 1 * 10 µG
Catalog # BIRBORB81388-10
Supplier: Biorbyt