ContactUsLinkComponent
sRAGE Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 134 amino acids and having a molecular mass of 15kDa. The Human sRAGE is fused to His tag at N-Terminus.The Human sRAGE is purified by pr 1 * 2 µG
  BIRBORB81387-2
 :  Biorbyt
sRAGE Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 134 amino acids and having a molecular mass of 15kDa. The Human sRAGE is fused to His tag at N-Terminus.The Human sRAGE is purified by pr 1 * 2 µG
  BIRBORB81387-2
 :  Biorbyt