ContactUsLinkComponent
SOX9 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 195 amino acids (316-510 a.a.) fused to GST tag and having total molecular mass of 48kDa. The SOX9 is purified by proprietary chromatograp 1 * 10 µG
Catalog # BIRBORB81379-10
Supplier:  Biorbyt
SOX9 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 195 amino acids (316-510 a.a.) fused to GST tag and having total molecular mass of 48kDa. The SOX9 is purified by proprietary chromatograp 1 * 10 µG
Catalog # BIRBORB81379-10
Supplier:  Biorbyt