ContactUsLinkComponent
SNAP25 Human Recombinant fused to N-terminal His-Tag produced in E.coli is a single, non-glycosylated polypeptide chain containing 2 26 amino acids and having a molecular mass of 25.4 kDa. 1 * 10 µG
Catalog # BIRBORB81370-10
Supplier:  Biorbyt
SNAP25 Human Recombinant fused to N-terminal His-Tag produced in E.coli is a single, non-glycosylated polypeptide chain containing 2 26 amino acids and having a molecular mass of 25.4 kDa. 1 * 10 µG
Catalog # BIRBORB81370-10
Supplier:  Biorbyt