ContactUsLinkComponent
SIRT6 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 375 amino acids (1-355 a.a.) a nd having a molecular mass of 41 kDa. Recombinant SIRT6 is fused to 20 amino acids His-tag at N-terminus and 1 * 2 µG
Catalog # BIRBORB81362-2
Supplier:  Biorbyt
SIRT6 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 375 amino acids (1-355 a.a.) a nd having a molecular mass of 41 kDa. Recombinant SIRT6 is fused to 20 amino acids His-tag at N-terminus and 1 * 2 µG
Catalog # BIRBORB81362-2
Supplier:  Biorbyt