ContactUsLinkComponent
SCGN Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 296 amino acids (1-276 a.a.) an d having a molecular mass of 34.2kDa. SCGN is fused to 20 amino acid His tag at N-terminus and purified by s 1 * 10 µG
Catalog # BIRBORB81352-10
Supplier:  Biorbyt
SCGN Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 296 amino acids (1-276 a.a.) an d having a molecular mass of 34.2kDa. SCGN is fused to 20 amino acid His tag at N-terminus and purified by s 1 * 10 µG
Catalog # BIRBORB81352-10
Supplier:  Biorbyt