ContactUsLinkComponent
S100B Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain fused to GST tag and having total molecu lar mass of 36kDa. The S100B is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81343-5
Supplier:  Biorbyt
S100B Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain fused to GST tag and having total molecu lar mass of 36kDa. The S100B is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81343-5
Supplier:  Biorbyt