ContactUsLinkComponent
S100B Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide containing 91 amino acids fragment (2-92) wit h an amino-terminal hexahistidine tag and having total molecular mass of 14.7kDa. The S100B is purified by p 1 * 5 µG
Catalog # BIRBORB81342-5
Supplier:  Biorbyt
S100B Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide containing 91 amino acids fragment (2-92) wit h an amino-terminal hexahistidine tag and having total molecular mass of 14.7kDa. The S100B is purified by p 1 * 5 µG
Catalog # BIRBORB81342-5
Supplier:  Biorbyt