S100A10 Human Recombinant also called Calpactin light chain is expressed in E.coli having molecular weight of 15.3kDa fused to an am ino terminal hexahistidine tag. 1 * 10 µG
Catalog # BIRBORB81340-10
Supplier: Biorbyt