ContactUsLinkComponent
CEA Human Recombinant is glycosylated with N-linked sugars and produced using baculovirus vectors in insect cells. CEA is well-known tumor marker corresponding to the full length human CEA which is approximately 120,000 dalton. 1 * 2 µG
Catalog # BIRBORB81020-2
Supplier:  Biorbyt
CEA Human Recombinant is glycosylated with N-linked sugars and produced using baculovirus vectors in insect cells. CEA is well-known tumor marker corresponding to the full length human CEA which is approximately 120,000 dalton. 1 * 2 µG
Catalog # BIRBORB81020-2
Supplier:  Biorbyt