CDC37 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 378 amino acids and having a m olecular mass of 44.4 kDa. 1 * 25 µG
Catalog # BIRBORB81018-25
Supplier: Biorbyt