ContactUsLinkComponent
HIV-1 TAT Cys22 Recombinant- produced in E.coli is a single, non-glycosylated, polypeptide chain containing 86 amino acids mutated i n the trans activation domain and having chain having a molecular mass of 14.4 kDa. 1 * 100 µG
Catalog # BIRBORB81727-100
Supplier:  Biorbyt
HIV-1 TAT Cys22 Recombinant- produced in E.coli is a single, non-glycosylated, polypeptide chain containing 86 amino acids mutated i n the trans activation domain and having chain having a molecular mass of 14.4 kDa. 1 * 100 µG
Catalog # BIRBORB81727-100
Supplier:  Biorbyt