ContactUsLinkComponent
CD2 Human Recombinant (aa 25-209) expressed in E.coli, shows 46 kDa band on SDS-PAGE.The CD2 is purified by proprietary chromatograp hic techniques. 1 * 2 µG
Catalog # BIRBORB81004-2
Supplier:  Biorbyt
CD2 Human Recombinant (aa 25-209) expressed in E.coli, shows 46 kDa band on SDS-PAGE.The CD2 is purified by proprietary chromatograp hic techniques. 1 * 2 µG
Catalog # BIRBORB81004-2
Supplier:  Biorbyt