ContactUsLinkComponent
CAPN1 consists of an 80-kDa large subunit and 30 kDa small subunit. CAPN1 was purified by sequential chromatography through DEAE-Sep harose, A1.5m Bio-Gel, and Phenyl-Sepharose CL-4B columns. 1 * 10 µG
Catalog # BIRBORB80996-10
Supplier:  Biorbyt
CAPN1 consists of an 80-kDa large subunit and 30 kDa small subunit. CAPN1 was purified by sequential chromatography through DEAE-Sep harose, A1.5m Bio-Gel, and Phenyl-Sepharose CL-4B columns. 1 * 10 µG
Catalog # BIRBORB80996-10
Supplier:  Biorbyt