ContactUsLinkComponent
Recombinant CALM2 produced in E.coli is a single, non-glycosylated polypeptide chain containing 149 amino acids and having a molecul ar mass of 16 kDa. CALM2 is purified by conventional chromatography techniques. 1 * 25 µG
Catalog # BIRBORB80986-25
Supplier:  Biorbyt
Recombinant CALM2 produced in E.coli is a single, non-glycosylated polypeptide chain containing 149 amino acids and having a molecul ar mass of 16 kDa. CALM2 is purified by conventional chromatography techniques. 1 * 25 µG
Catalog # BIRBORB80986-25
Supplier:  Biorbyt