Recoverin Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 200 amino acids having a m olecular mass of 23kDa. 1 * 10 µG
Catalog # BIRBORB81311-10
Supplier: Biorbyt