ContactUsLinkComponent
Retinoblastoma Human Recombinant fused with 6X His tag produced in E.coli is a single, non-glycosylated, polypeptide chain containin g 146 amino acids and having a molecular mass of 16.5 kDa.The Retinoblastoma is purified by proprietary chro 1 * 10 µG
Catalog # BIRBORB81308-10
Supplier:  Biorbyt
Retinoblastoma Human Recombinant fused with 6X His tag produced in E.coli is a single, non-glycosylated, polypeptide chain containin g 146 amino acids and having a molecular mass of 16.5 kDa.The Retinoblastoma is purified by proprietary chro 1 * 10 µG
Catalog # BIRBORB81308-10
Supplier:  Biorbyt