ContactUsLinkComponent
Recombinant RAC2 produced in E.coli is a single, non-glycosylated polypeptide chain containing 419 amino acids and having a molecula r mass of 47 kDa. RAC2 is fused to GST and purified by conventional chromatography techniques. 1 * 20 µG
Catalog # BIRBORB81300-20
Supplier:  Biorbyt
Recombinant RAC2 produced in E.coli is a single, non-glycosylated polypeptide chain containing 419 amino acids and having a molecula r mass of 47 kDa. RAC2 is fused to GST and purified by conventional chromatography techniques. 1 * 20 µG
Catalog # BIRBORB81300-20
Supplier:  Biorbyt