ContactUsLinkComponent
Ras-Related C3 Botulinum Toxin substrate Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain contai ning 192 amino acids and having a molecular mass of 21.4 kDa. 1 * 10 µG
Catalog # BIRBORB81298-10
Supplier:  Biorbyt
Ras-Related C3 Botulinum Toxin substrate Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain contai ning 192 amino acids and having a molecular mass of 21.4 kDa. 1 * 10 µG
Catalog # BIRBORB81298-10
Supplier:  Biorbyt