ContactUsLinkComponent
PSP94 Recombinant Human produced in E.coli is a single, non-glycosylated, polypeptide chain containing 104 amino acids and having a molecular mass of 12 kDa. The PSP94 is fused to His tag at N-Terminus.The Human IGBF is purified by propriet 1 * 2 µG
Catalog # BIRBORB81288-2
Supplier:  Biorbyt
PSP94 Recombinant Human produced in E.coli is a single, non-glycosylated, polypeptide chain containing 104 amino acids and having a molecular mass of 12 kDa. The PSP94 is fused to His tag at N-Terminus.The Human IGBF is purified by propriet 1 * 2 µG
Catalog # BIRBORB81288-2
Supplier:  Biorbyt