ContactUsLinkComponent
Survivin Human Recombinant fused to 152 a.a. N-terminal CaM-Tag produced in E.coli is a single, non-glycosylated polypeptide chain c ontaining 294 amino acids (1-142 a.a.) and having a molecular mass of 33 kDa. 1 * 25 µG
Catalog # BIRBORB80967-25
Supplier:  Biorbyt
Survivin Human Recombinant fused to 152 a.a. N-terminal CaM-Tag produced in E.coli is a single, non-glycosylated polypeptide chain c ontaining 294 amino acids (1-142 a.a.) and having a molecular mass of 33 kDa. 1 * 25 µG
Catalog # BIRBORB80967-25
Supplier:  Biorbyt