ContactUsLinkComponent
YWHAG Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 247 amino acids (1-247) and h aving a molecular mass of 28 kDa. YWHAG is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80881-5
Supplier:  Biorbyt
YWHAG Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 247 amino acids (1-247) and h aving a molecular mass of 28 kDa. YWHAG is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80881-5
Supplier:  Biorbyt