PKM2 Human Recombinant is produced in E.coli is a single, non-glycosylated, polypeptide having a molecular mass of 62400 Dalton wit h an amino-terminal hexahistidine tag. The PKM2 is purified by standard chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80859-10
Supplier: Biorbyt