ContactUsLinkComponent
PDK1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 429 amino acids (29-436) and h aving a molecular mass of 48.6 kDa. PDK1 is fused to 20 a.a. His-Tag at N-terminus and purified by proprieta 1 * 10 µG
Catalog # BIRBORB80840-10
Supplier:  Biorbyt
PDK1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 429 amino acids (29-436) and h aving a molecular mass of 48.6 kDa. PDK1 is fused to 20 a.a. His-Tag at N-terminus and purified by proprieta 1 * 10 µG
Catalog # BIRBORB80840-10
Supplier:  Biorbyt