ContactUsLinkComponent
NME2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 152 amino acids (1-152 a.a.) an d having a molecular mass of 17.2kDa. The NME2 is purified by conventional chromatography. 1 * 25 µG
Catalog # BIRBORB80835-25
Supplier:  Biorbyt
NME2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 152 amino acids (1-152 a.a.) an d having a molecular mass of 17.2kDa. The NME2 is purified by conventional chromatography. 1 * 25 µG
Catalog # BIRBORB80835-25
Supplier:  Biorbyt