ContactUsLinkComponent
Soluble LYVE1 Mouse Recombinant fused to C-terminal His-tag (6xHis) produced in baculovirus is monomeric, glycosylated, polypeptide containing 228 amino acids (Met-1 to Gly 228) and having a molecular mass of 25 kDa but as result of glycosi 1 * 2 µG
  BIRBORB80832-2
 :  Biorbyt
Soluble LYVE1 Mouse Recombinant fused to C-terminal His-tag (6xHis) produced in baculovirus is monomeric, glycosylated, polypeptide containing 228 amino acids (Met-1 to Gly 228) and having a molecular mass of 25 kDa but as result of glycosi 1 * 2 µG
  BIRBORB80832-2
 :  Biorbyt