ContactUsLinkComponent
LYVE1 Human Recombinant produced in insect cells is a single, glycosylated polypeptide chain containing 229 amino acids and having a molecular mass of 24.8 kDa. As result of glycosylation, the LYVE1 migrates on SDS-PAGE at approximately 50 1 * 10 µG
Catalog # BIRBORB80830-10
Supplier:  Biorbyt
LYVE1 Human Recombinant produced in insect cells is a single, glycosylated polypeptide chain containing 229 amino acids and having a molecular mass of 24.8 kDa. As result of glycosylation, the LYVE1 migrates on SDS-PAGE at approximately 50 1 * 10 µG
Catalog # BIRBORB80830-10
Supplier:  Biorbyt