ContactUsLinkComponent
HK-3 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain fused to His tag at the N-terminal encodi ng the sequence of 943 amino acids and having a molecular mass of 101.1 kDa.HXK3 is purified by proprietary 1 * 2 µG
Catalog # BIRBORB80822-2
Supplier:  Biorbyt
HK-3 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain fused to His tag at the N-terminal encodi ng the sequence of 943 amino acids and having a molecular mass of 101.1 kDa.HXK3 is purified by proprietary 1 * 2 µG
Catalog # BIRBORB80822-2
Supplier:  Biorbyt