ContactUsLinkComponent
GUK1 Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 217 amino acids (1-197 a.a.)and having total molecular mass of 23.9 kDa. GUK1 is fused to 20 amino acid His Tag at N-terminus and is purifie 1 * 20 µG
Catalog # BIRBORB80819-20
Supplier:  Biorbyt
GUK1 Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 217 amino acids (1-197 a.a.)and having total molecular mass of 23.9 kDa. GUK1 is fused to 20 amino acid His Tag at N-terminus and is purifie 1 * 20 µG
Catalog # BIRBORB80819-20
Supplier:  Biorbyt