ContactUsLinkComponent
IL-29 human recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 181 amino acids and having a molecular mass of 20 kDa. 1 * 5 µG
Catalog # BIRBORB80476-5
Supplier:  Biorbyt
IL-29 human recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 181 amino acids and having a molecular mass of 20 kDa. 1 * 5 µG
Catalog # BIRBORB80476-5
Supplier:  Biorbyt