ContactUsLinkComponent
IL27 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 207 amino acids and having a mo lecular mass of 23.7 kDa. The Murine IL-27 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80474-2
Supplier:  Biorbyt
IL27 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 207 amino acids and having a mo lecular mass of 23.7 kDa. The Murine IL-27 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80474-2
Supplier:  Biorbyt