IL22 Mouse Recombinant produced in E.coli is a single, non-glycosylated homodimeric polypeptide chain containing 147 amino acids and having a molecular mass of 16.7 kDa. The Murine IL-22 is purified by proprietary chromatographic techniques 1 * 10 µG
Catalog # BIRBORB80471-10
Supplier: Biorbyt