IL20 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 152 amino acids and having tota l molecular mass of 17,584 Dalton. The IL-20 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80469-10
Supplier: Biorbyt