ContactUsLinkComponent
IL19 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 155 amino acids and having a mo lecular mass of 17913 Dalton. The IL-19 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80467-2
Supplier:  Biorbyt
IL19 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 155 amino acids and having a mo lecular mass of 17913 Dalton. The IL-19 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80467-2
Supplier:  Biorbyt