BMP2 Human Recombinant produced in E.coli is monomeric, non-glycosylated, Polypeptide chain containing 115 amino acids (283-396) and having a molecular mass of 13009 Dalton. The BMP-2 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80274-20
Supplier: Biorbyt