BMPR1A Human Recombinant produced in HEK-293 cells is a single, glycosylated, polypeptide chain containing 369 amino acids with mole cular weight of 43.7 kDa though on SDS-PAGE migrates at about 50 kDa due to the glycosilation. The BMPR1A is 1 * 2 µG
Catalog # BIRBORB80273-2
Supplier: Biorbyt