ContactUsLinkComponent
IL17E Rat Recombinant produced in E.coli is homodimeric, non-glycosylated polypeptide chain containing 145 amino acids and having a molecular mass of 35.5 kDa. The IL-25 Rat is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80461-5
Supplier:  Biorbyt
IL17E Rat Recombinant produced in E.coli is homodimeric, non-glycosylated polypeptide chain containing 145 amino acids and having a molecular mass of 35.5 kDa. The IL-25 Rat is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80461-5
Supplier:  Biorbyt