ContactUsLinkComponent
IL17 A/F Mouse Recombinant produced in E.coli is heterodimeric, non-glycosylated polypeptide comprised of IL17A monomeric subunit an d IL17F monomeric subunit containing total of 266 amino acids and having total molecular mass of 29.8kDa. Th 1 * 100 µG
Catalog # BIRBORB80458-100
Supplier:  Biorbyt
IL17 A/F Mouse Recombinant produced in E.coli is heterodimeric, non-glycosylated polypeptide comprised of IL17A monomeric subunit an d IL17F monomeric subunit containing total of 266 amino acids and having total molecular mass of 29.8kDa. Th 1 * 100 µG
Catalog # BIRBORB80458-100
Supplier:  Biorbyt