ContactUsLinkComponent
IL17E Mouse Recombinant produced in E.coli is homodimeric, non-glycosylated polypeptide chain containing total of 306 amino acids an d having a molecular mass of 34.9 kDa. The Mouse IL-17E is purified by proprietary chromatographic technique 1 * 25 µG
Catalog # BIRBORB80456-25
Supplier:  Biorbyt
IL17E Mouse Recombinant produced in E.coli is homodimeric, non-glycosylated polypeptide chain containing total of 306 amino acids an d having a molecular mass of 34.9 kDa. The Mouse IL-17E is purified by proprietary chromatographic technique 1 * 25 µG
Catalog # BIRBORB80456-25
Supplier:  Biorbyt