ContactUsLinkComponent
Glucagon Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 29 amino acids and having a molecular mass of 3483 Dalton. The Glucagon is purified by proprietary chromatographic techniques. 1 * 1 mg
Catalog # BIRBORB80726-1
Supplier:  Biorbyt
Glucagon Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 29 amino acids and having a molecular mass of 3483 Dalton. The Glucagon is purified by proprietary chromatographic techniques. 1 * 1 mg
Catalog # BIRBORB80726-1
Supplier:  Biorbyt