ContactUsLinkComponent
Growth Hormone Releasing Peptide-6 Synthetic is a single, non-glycosylated polypeptide chain containing amino acids, having a molecu lar mass of 873 Dalton and a molecular formula of C46H56N12O6. 1 * 25 mg
Catalog # BIRBORB80721-25
Supplier:  Biorbyt
Growth Hormone Releasing Peptide-6 Synthetic is a single, non-glycosylated polypeptide chain containing amino acids, having a molecu lar mass of 873 Dalton and a molecular formula of C46H56N12O6. 1 * 25 mg
Catalog # BIRBORB80721-25
Supplier:  Biorbyt