IL15 His Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 123 amino acids fragment ( 49-162) having total molecular weight of 13.9 kDa with an C-terminal hexahistidine tag. The IL-15 His is pur 1 * 2 µG
Catalog # BIRBORB80444-2
Supplier: Biorbyt