ContactUsLinkComponent
The AIF1 Human Recombinant contains total of 155 amino acids having a molecular mass of 17.7kDa. The Human AIF1 is fused to 9 amino acid long N-terminal His tag. 1 * 10 µG
Catalog # BIRBORB80254-10
Supplier:  Biorbyt
The AIF1 Human Recombinant contains total of 155 amino acids having a molecular mass of 17.7kDa. The Human AIF1 is fused to 9 amino acid long N-terminal His tag. 1 * 10 µG
Catalog # BIRBORB80254-10
Supplier:  Biorbyt