ContactUsLinkComponent
Growth Hormone Releasing Peptide-2 Synthetic is a single, non-glycosylated polypeptide chain containing amino acids, having a molecu lar mass of 746.9 Dalton and a molecular formula of C42H50N8O5. 1 * 25 mg
Catalog # BIRBORB80720-25
Supplier:  Biorbyt
Growth Hormone Releasing Peptide-2 Synthetic is a single, non-glycosylated polypeptide chain containing amino acids, having a molecu lar mass of 746.9 Dalton and a molecular formula of C42H50N8O5. 1 * 25 mg
Catalog # BIRBORB80720-25
Supplier:  Biorbyt