ContactUsLinkComponent
Ghrelin Human contains 28 amino acids and total molecular mass of 3370.9 Dalton and a molecular formula of C149H249N47O42.The GHRL i s purified by proprietary chromatographic techniques. 1 * 1 mg
Catalog # BIRBORB80718-1
Supplier:  Biorbyt
Ghrelin Human contains 28 amino acids and total molecular mass of 3370.9 Dalton and a molecular formula of C149H249N47O42.The GHRL i s purified by proprietary chromatographic techniques. 1 * 1 mg
Catalog # BIRBORB80718-1
Supplier:  Biorbyt