ContactUsLinkComponent
Growth Hormone Releasing Hormone Human Synthetic is a single, non-glycosylated, polypeptide chain containing 29 amino acids and havi ng a molecular mass of 3358.9 Dalton.Corresponds to the amino-terminal segment of the naturally occurring hu 1 * 250 µG
Catalog # BIRBORB80717-250
Supplier:  Biorbyt
Growth Hormone Releasing Hormone Human Synthetic is a single, non-glycosylated, polypeptide chain containing 29 amino acids and havi ng a molecular mass of 3358.9 Dalton.Corresponds to the amino-terminal segment of the naturally occurring hu 1 * 250 µG
Catalog # BIRBORB80717-250
Supplier:  Biorbyt