ContactUsLinkComponent
Soluble TNF-related apoptosis-inducing ligand Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain c ontaining 168 amino acids and having a molecular mass of 19.6 kDa.The sTRAIL is purified by proprietary chro 1 * 50 µG
Catalog # BIRBORB80629-50
Supplier:  Biorbyt
Soluble TNF-related apoptosis-inducing ligand Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain c ontaining 168 amino acids and having a molecular mass of 19.6 kDa.The sTRAIL is purified by proprietary chro 1 * 50 µG
Catalog # BIRBORB80629-50
Supplier:  Biorbyt