ContactUsLinkComponent
TNFR1 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 161 amino acids fragment (41- 201) having molecular weight of 22.68kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The TNFR1, 1 * 5 µG
Catalog # BIRBORB80624-5
Supplier:  Biorbyt
TNFR1 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 161 amino acids fragment (41- 201) having molecular weight of 22.68kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The TNFR1, 1 * 5 µG
Catalog # BIRBORB80624-5
Supplier:  Biorbyt