ContactUsLinkComponent
IL10 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 181 amino acids fragment (19-1 78) and having total molecular mass of 21 kDa with 20 amino acids N-Terminal hexahistidine tag. The IL-10 Hi 1 * 2 µG
Catalog # BIRBORB80437-2
Supplier:  Biorbyt
IL10 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 181 amino acids fragment (19-1 78) and having total molecular mass of 21 kDa with 20 amino acids N-Terminal hexahistidine tag. The IL-10 Hi 1 * 2 µG
Catalog # BIRBORB80437-2
Supplier:  Biorbyt